Assay Details
Target Gene Details
Entrez Gene ID: | 66153 |
Gene Name: | F-box protein 36 |
Gene Aliases: |
0610008D19Rik, 1110020F21Rik, 2410002G19Rik, D1Ertd757e |
Location: |
Chr.1:84839841-84900487 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxo36 | NM_025386.3 | NP_079662.1 | ||
| AK003856.1 | BAB23040.1 | |||
| AK008324.1 | BAB25602.1 | |||
| AK036072.1 | ||||
| BC049581.1 | AAH49581.1 | |||
| BY096703.1 | ||||
| BY708252.1 |
Target Gene Details
Entrez Gene ID: | 14897 |
Gene Name: | thyroid hormone receptor interactor 12 |
Gene Aliases: |
1110036I07Rik, 6720416K24Rik, AA410158, Gtl6, TRIP-12 |
Location: |
Chr.1:84721189-84850008 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trip12 | AK134397.1 | BAE22126.1 | ||
| AK164719.1 | BAE37889.1 |
More Information
Set Membership: |
|

Genomic Map