Assay Details
Target Gene Details
Entrez Gene ID: | 64294 |
Gene Name: | integral membrane protein 2C |
Gene Aliases: |
3110038L02Rik, BRI3, Bricd2c, E25, E25C, ITM3 |
Location: |
Chr.1:85894510-85908698 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itm2c | NM_022417.3 | 3 | 553 | NP_071862.2 |
| AB030199.1 | 3 | 496 | BAA92762.1 | |
| AF272044.1 | 3 | 460 | AAF89493.1 | |
| AF282981.1 | 3 | 481 | AAK00277.1 | |
| AK036377.1 | 3 | 499 | BAC29403.1 | |
| AK156930.1 | 3 | 456 | BAE33901.1 | |
| AK194904.1 | ||||
| AK207985.1 | ||||
| BC012952.1 | 3 | 431 | AAH12952.1 |

Genomic Map