Assay Details
Target Gene Details
Entrez Gene ID: | 64294 |
Gene Name: | integral membrane protein 2C |
Gene Aliases: |
3110038L02Rik, BRI3, Bricd2c, E25, E25C, ITM3 |
Location: |
Chr.1:85894510-85908698 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itm2c | NM_022417.3 | 5 | 797 | NP_071862.2 |
| AB030199.1 | 5 | 740 | BAA92762.1 | |
| AF272044.1 | 5 | 704 | AAF89493.1 | |
| AF282981.1 | 5 | 725 | AAK00277.1 | |
| AK014144.1 | ||||
| AK036377.1 | 5 | 743 | BAC29403.1 | |
| AK156930.1 | 5 | 700 | BAE33901.1 | |
| BC012952.1 | 5 | 675 | AAH12952.1 |

Genomic Map