Assay Details
Target Gene Details
Entrez Gene ID: | 75605 |
Gene Name: | lysine (K)-specific demethylase 5B |
Gene Aliases: |
2010009J12Rik, 2210016I17Rik, AW556288, D1Ertd202e, Jarid1b, PLU-1, PUT1, Plu1, RBBP2H1A, Rb-Bp2, mKIAA4034 |
Location: |
Chr.1:134560178-134632883 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kdm5b | NM_152895.2 | 12 | 1776 | NP_690855.2 |
| AK041304.1 | 12 | 1783 | BAC30898.1 | |
| AK163481.1 | 12 | 1739 | BAE37363.1 | |
| AK220451.1 | 12 | 1762 | BAD90482.1 | |
| AY082429.1 | 12 | 1759 | AAL92848.1 | |
| AY082430.1 | 12 | 1759 | AAL92849.1 | |
| BC048180.1 | 12 | 1759 | AAH48180.1 | |
| BC057318.1 | 12 | 1759 | AAH57318.1 |
More Information
Set Membership: |
|

Genomic Map