Assay Details
Target Gene Details
Entrez Gene ID: | 68421 |
Gene Name: | LMBR1 domain containing 1 |
Gene Aliases: |
0910001K20Rik, AV347960 |
Location: |
Chr.1:24678544-24766301 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmbrd1 | NM_026719.2 | 9 | 1041 | NP_080995.2 |
| AK152170.1 | 9 | 852 | BAE31002.1 | |
| AK153237.1 | 9 | 852 | BAE31829.1 | |
| AK158982.1 | 9 | 877 | BAE34756.1 | |
| AK167080.1 | 9 | 860 | BAE39240.1 | |
| AY061989.1 | 8 | 752 | AAL38052.1 | |
| BC025579.1 | 9 | 873 | AAH25579.1 | |
| BC027774.1 | 9 | 924 | AAH27774.1 | |
| BC031902.1 | 9 | 864 | AAH31902.1 | |
| BC039053.1 | 9 | 955 | AAH39053.1 |
More Information
Set Membership: |
|

Genomic Map