Assay Details
Target Gene Details
Entrez Gene ID: | 16565 |
Gene Name: | kinesin family member 21B |
Gene Aliases: |
2610511N21Rik, mKIAA0449 |
Location: |
Chr.1:136131374-136178014 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif21b | NM_001039472.1 | NP_001034561.1 | ||
| AF202893.1 | AAF17084.1 |
More Information
Set Membership: |
|

Genomic Map