Assay Details
Target Gene Details
Entrez Gene ID: | 16565 |
Gene Name: | kinesin family member 21B |
Gene Aliases: |
2610511N21Rik, mKIAA0449 |
Location: |
Chr.1:136131374-136178014 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif21b | NM_001039472.1 | 8 | 1477 | NP_001034561.1 |
| AF202893.1 | 8 | 1455 | AAF17084.1 | |
| AK089400.1 | 6 | 2011 | BAC40866.1 |
More Information
Set Membership: |
|

Genomic Map