Assay Details
Target Gene Details
Entrez Gene ID: | 213417 |
Gene Name: | kelch domain containing 8A |
Gene Aliases: |
A630065K24Rik |
Location: |
Chr.1:132298490-132307357 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhdc8a | NM_144810.5 | 4 | 1465 | NP_659059.1 |
| AK043039.1 | 4 | 803 | BAC31445.1 | |
| AK044318.1 | 4 | 1041 | BAC31864.1 | |
| AK080356.1 | 4 | 1465 | BAC37888.1 | |
| BC011135.1 | 4 | 1030 | AAH11135.1 | |
| BC023173.1 | 4 | 664 | AAH23173.1 |

Genomic Map