Assay Details
Target Gene Details
Entrez Gene ID: | 17528 |
Gene Name: | myelin protein zero |
Gene Aliases: |
Mpp, P-zero, P0 |
Location: |
Chr.1:171150713-171161125 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpz | NM_008623.4 | 4 | 317 | NP_032649.2 |
| AK089180.1 | 4 | 321 | BAC40781.1 | |
| AK157960.1 | 2 | 214 | BAE34285.1 | |
| BC139139.1 | 2 | 182 | ||
| BC141226.1 | 2 | 182 | ||
| BY201877.1 | 4 | 321 | ||
| BY339244.1 | 2 | 207 |
More Information
Set Membership: |
|

Genomic Map