Assay Details
Target Gene Details
Entrez Gene ID: | 68481 |
Gene Name: | myelin protein zero-like 1 |
Gene Aliases: |
1110007A10Rik, PZR |
Location: |
Chr.1:165592181-165634558 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpzl1 | NM_001001880.2 | NP_001001880.1 | ||
| NM_001083897.1 | NP_001077366.1 | |||
| AK076882.1 | ||||
| AK090278.1 | ||||
| AK169385.1 | BAE41132.1 | |||
| AK189245.1 | ||||
| AY764247.1 | AAX11678.1 | |||
| AY764248.1 | AAX11679.1 | |||
| BC072563.1 | AAH72563.1 |
More Information
Set Membership: |
|

Genomic Map