Assay Details
Target Gene Details
Entrez Gene ID: | 227197 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 1 |
Gene Aliases: |
5830412M15Rik, 9930026A05Rik |
Location: |
Chr.1:63143592-63176822 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufs1 | NM_001160038.1 | 5 | 713 | NP_001153510.1 |
| NM_001160039.1 | 5 | 555 | NP_001153511.1 | |
| NM_001160040.1 | 4 | 609 | NP_001153512.1 | |
| NM_145518.2 | 4 | 451 | NP_663493.2 | |
| AK034597.1 | 4 | 520 | BAE20494.1 | |
| AK036926.1 | 4 | 316 | BAC29641.1 | |
| AK078590.1 | 4 | 297 | ||
| AK142711.1 | 4 | 331 | BAE25170.1 | |
| AK146814.1 | 4 | 469 | BAE27454.1 | |
| AK166604.1 | 5 | 402 | ||
| AK167705.1 | 4 | 299 | BAE39749.1 | |
| AK178076.1 | 3 | 221 | ||
| AK180817.1 | 2 | 100 | ||
| AK213867.1 | 1 | 61 | ||
| BC006660.1 | 4 | 338 | AAH06660.1 | |
| BC015300.1 | 4 | 271 | AAH15300.1 | |
| BG808119.1 | 4 | 449 | ||
| BU708393.1 | 5 | 556 | ||
| BY709888.1 | 4 | 452 | ||
| CN715066.1 | 4 | 327 |
More Information
Set Membership: |
|

Genomic Map