Assay Details
Target Gene Details
Entrez Gene ID: | 18627 |
Gene Name: | period circadian clock 2 |
Gene Aliases: |
mKIAA0347, mPer2 |
Location: |
Chr.1:91415979-91459393 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Per2 | NM_011066.3 | 12 | 1538 | NP_035196.2 |
| AF035830.1 | 12 | 1485 | AAC53592.1 | |
| AF036893.1 | 12 | 1508 | AAC39942.1 | |
| AK044658.1 | 12 | 1483 | BAC32022.1 | |
| AK122253.1 | 12 | 1503 | BAC65535.1 | |
| AK159847.1 | 12 | 1540 | BAE35425.1 | |
| AK165556.1 | 12 | 1541 | BAE38253.1 |
More Information
Set Membership: |
|

Genomic Map