Assay Details
Target Gene Details
Entrez Gene ID: | 18484 |
Gene Name: | peptidylglycine alpha-amidating monooxygenase |
Gene Aliases: |
PHM |
Location: |
Chr.1:97821091-98095632 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pam | NM_013626.3 | 21 | 2729 | NP_038654.2 |
| AK132581.1 | 9 | 1155 | BAE21243.1 | |
| BC129869.1 | 4 | 409 | ||
| U79523.1 | 21 | 2614 | AAB38364.1 |
More Information
Set Membership: |
|

Genomic Map