Assay Details
Target Gene Details
Entrez Gene ID: | 240752 |
Gene Name: | phosphoinositide-3-kinase, class 2, beta polypeptide |
Gene Aliases: |
C330011J12Rik, Gm199, PI3K-C2beta, PI3KC2beta |
Location: |
Chr.1:133045672-133108688 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3c2b | NM_001099276.2 | NP_001092746.2 |
More Information
Set Membership: |
|

Genomic Map