Assay Details
Target Gene Details
Entrez Gene ID: | 102465544 |
Gene Name: | microRNA 6902 |
Gene Aliases: |
mmu-mir-6902 |
Location: |
Chr.1:93346092-93346155 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir6902 | NR_105867.1 |
Target Gene Details
Entrez Gene ID: | 66385 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 7 |
Gene Aliases: |
2310014J01Rik, SDS22 |
Location: |
Chr.1:93343645-93367619 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r7 | NM_023200.2 | NP_075689.1 | ||
| AF222867.1 | AAK00624.1 | |||
| AK009342.1 | ||||
| AK009870.1 | BAB26554.1 | |||
| AK129024.1 | ||||
| AK145136.1 | BAE26253.1 | |||
| AK162517.1 | BAE36954.1 | |||
| BC013524.1 | AAH13524.1 |
More Information
Set Membership: |
|

Genomic Map