Assay Details
Target Gene Details
Entrez Gene ID: | 19076 |
Gene Name: | DNA primase, p58 subunit |
Gene Aliases: |
AI323589 |
Location: |
Chr.1:33453808-33669794 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prim2 | NM_008922.2 | NP_032948.1 | ||
| AK076095.1 | BAC36179.1 | |||
| AK077760.1 | BAC36996.1 | |||
| AK153970.1 | BAE32287.1 | |||
| AK166687.1 | BAE38946.1 | |||
| AK168246.1 | BAE40196.1 | |||
| BC019500.1 | AAH19500.1 | |||
| D13545.1 | BAA02745.1 | |||
| D17385.1 | BAA04203.1 |
More Information
Set Membership: |
|

Genomic Map