Assay Details
Target Gene Details
Entrez Gene ID: | 19076 |
Gene Name: | DNA primase, p58 subunit |
Gene Aliases: |
AI323589 |
Location: |
Chr.1:33453808-33669794 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prim2 | NM_008922.2 | 2 | 259 | NP_032948.1 |
| AK076095.1 | 2 | 241 | BAC36179.1 | |
| AK077760.1 | 2 | 246 | BAC36996.1 | |
| AK153970.1 | 2 | 220 | BAE32287.1 | |
| AK168246.1 | 2 | 252 | BAE40196.1 | |
| BC019500.1 | 2 | 220 | AAH19500.1 | |
| BY131806.1 | 2 | 260 | ||
| D13545.1 | 2 | 223 | BAA02745.1 | |
| D17385.1 | 2 | 203 | BAA04203.1 |
More Information
Set Membership: |
|

Genomic Map