Assay Details
Target Gene Details
Entrez Gene ID: | 19264 |
Gene Name: | protein tyrosine phosphatase, receptor type, C |
Gene Aliases: |
B220, CD45R, Cd45, L-CA, Ly-5, Lyt-4, T200, loc |
Location: |
Chr.1:138062859-138175755 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprc | NM_001111316.2 | 30 | 3474 | NP_001104786.2 |
| NM_001268286.1 | 27 | 3083 | NP_001255215.1 | |
| NM_011210.4 | 28 | 3155 | NP_035340.3 | |
| AK154893.1 | 27 | 3078 | BAE32907.1 | |
| CA575153.1 | 4 | 495 | ||
| CA577897.1 | 1 | 134 | ||
| M14342.1 | 28 | 3117 | AAA39458.1 | |
| M14343.1 | 24 | 3648 | ||
| M15174.1 | 5 | 550 | AAA40161.1 | |
| M92933.1 | 30 | 3474 | AAA39459.1 |
More Information
Set Membership: |
|

Genomic Map