Assay Details
Target Gene Details
Entrez Gene ID: | 98711 |
Gene Name: | retinol dehydrogenase 10 (all-trans) |
Gene Aliases: |
3110069K09Rik, 4921506A21Rik, AI875664, AW549993, D1Ertd762e, m366Asp |
Location: |
Chr.1:16105780-16133734 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rdh10 | NM_133832.3 | NP_598593.1 | ||
| AF456767.1 | AAN64749.1 | |||
| AK149656.1 | ||||
| BC019796.1 | AAH19796.2 |

Genomic Map