Assay Details
Target Gene Details
Entrez Gene ID: | 109232 |
Gene Name: | saccharopine dehydrogenase (putative) |
Gene Aliases: |
AW214504, C330023F11Rik |
Location: |
Chr.1:179668231-179687184 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sccpdh | NM_178653.3 | NP_848768.1 | ||
| AK042919.1 | BAC31405.1 | |||
| AK046033.1 | BAC32578.1 | |||
| AK046468.1 | ||||
| AK050554.1 | ||||
| AK077695.1 | BAC36962.1 | |||
| AK082803.1 | BAC38628.1 | |||
| AK086448.1 | ||||
| AK086632.1 | ||||
| AK145399.1 | BAE26412.1 | |||
| AK166018.1 | BAE38523.1 | |||
| AK212321.1 | ||||
| BC025803.1 | AAH25803.1 |

Genomic Map