Assay Details
Target Gene Details
Entrez Gene ID: | 74748 |
Gene Name: | SLAM family member 8 |
Gene Aliases: |
5830408F06Rik, Blame, SBBI42 |
Location: |
Chr.1:172581377-172591191 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slamf8 | NM_029084.3 | NP_083360.2 | ||
| AK142633.1 | BAE25140.1 |
More Information
Set Membership: |
|

Genomic Map