Assay Details
Target Gene Details
Entrez Gene ID: | 14270 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 2 |
Gene Aliases: |
9930124L22Rik, AI448945, FBP2, Fnbp2, srGAP3 |
Location: |
Chr.1:131285251-131527369 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srgap2 | NM_001081011.2 | 3 | 843 | NP_001074480.2 |
| AK084412.1 | 2 | 722 | ||
| AK143649.1 | 3 | 834 | BAE25478.1 | |
| BC151081.1 | 2 | 461 | ||
| BC151082.1 | 2 | 461 | ||
| BC158055.1 | 2 | 459 | ||
| BC172152.1 | 2 | 461 | ||
| CF164437.1 | 2 | 120 |
More Information
Set Membership: |
|

Genomic Map