Assay Details
Target Gene Details
Entrez Gene ID: | 226830 |
Gene Name: | SET and MYND domain containing 2 |
Gene Aliases: |
1110020E07Rik, 4930402C15, KMT3C, Zmynd14 |
Location: |
Chr.1:189880492-189922379 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smyd2 | NM_026796.1 | 3 | 308 | NP_081072.1 |
| AK145261.1 | 3 | 313 | ||
| AK150857.1 | 3 | 312 | BAE29912.1 | |
| AK164053.1 | 3 | 274 | ||
| AK178146.1 | ||||
| AK197062.1 | ||||
| BC023119.1 | 3 | 308 | AAH23119.1 |
More Information
Set Membership: |
|

Genomic Map