Assay Details
Target Gene Details
Entrez Gene ID: | 240690 |
Gene Name: | suppression of tumorigenicity 18 |
Gene Aliases: |
AV348974, Myt3, NZF-3, Nzf3, mKIAA0535 |
Location: |
Chr.1:6487231-6860940 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| St18 | NM_001244692.1 | 23 | 3347 | NP_001231621.1 |
| NM_001244693.1 | 22 | 3269 | NP_001231622.1 | |
| NM_173868.2 | 21 | 3160 | NP_776293.1 | |
| NR_045188.1 | 23 | 3311 | ||
| NR_045189.1 | 22 | 3233 | ||
| AB097467.1 | 21 | 3185 | ||
| AJ536155.1 | 23 | 3311 | CAD60089.1 | |
| AK122303.1 | 21 | 3109 | BAC65585.1 | |
| AK137440.1 | 21 | 3110 | BAE23351.1 | |
| AK147320.1 | 23 | 3348 | BAE27844.1 | |
| AK147664.1 | 23 | 3348 | BAE28057.1 | |
| BC032273.1 | 16 | 2074 | AAH32273.1 | |
| BC118528.1 | 22 | 3183 |
More Information
Set Membership: |
|

Genomic Map