Assay Details
Target Gene Details
Entrez Gene ID: | 74717 |
Gene Name: | spermatogenesis associated 17 |
Gene Aliases: |
1700065F16Rik, 4930504I07Rik, 4930513F16Rik, MSRG-11 |
Location: |
Chr.1:187009614-187215462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata17 | NM_028848.3 | NP_083124.1 |

Genomic Map