Assay Details
Target Gene Details
Entrez Gene ID: | 208795 |
Gene Name: | transmembrane protein 63a |
Gene Aliases: |
BC014795, Tmem64a |
Location: |
Chr.1:180941225-180975106 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem63a | NM_144794.2 | 13 | 1200 | NP_659043.1 |
| AK049360.1 | 13 | 1194 | ||
| AK144899.1 | 13 | 1253 | BAE26121.1 | |
| AK170559.1 | 13 | 1216 | BAE41880.1 | |
| AK173027.1 | 13 | 1193 | BAD32305.1 | |
| AK218532.1 | 2 | 127 | ||
| AK219713.1 | 2 | 127 | ||
| BC014795.1 | 13 | 1173 | AAH14795.1 | |
| BC019442.1 | 13 | 1200 | AAH19442.1 |

Genomic Map