Assay Details
Target Gene Details
Entrez Gene ID: | 208795 |
Gene Name: | transmembrane protein 63a |
Gene Aliases: |
BC014795, Tmem64a |
Location: |
Chr.1:180941225-180975106 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem63a | NM_144794.2 | NP_659043.1 | ||
AK049360.1 | ||||
AK144899.1 | BAE26121.1 | |||
AK170559.1 | BAE41880.1 | |||
AK173027.1 | BAD32305.1 | |||
AK215121.1 | ||||
BC014795.1 | AAH14795.1 | |||
BC019442.1 | AAH19442.1 |
