Assay Details
Target Gene Details
Entrez Gene ID: | 211484 |
Gene Name: | testis specific 10 |
Gene Aliases: |
4933432N21Rik, AI843383, Gm217, Mtsga10 |
Location: |
Chr.1:37728144-37866739 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tsga10 | NM_001290720.1 | 15 | 1983 | NP_001277649.1 |
| NM_001290721.1 | 14 | 1790 | NP_001277650.1 | |
| NM_207228.3 | 15 | 1850 | NP_997111.1 | |
| NR_110976.1 | 13 | 1635 | ||
| AF530050.3 | 14 | 1795 | AAN01136.3 | |
| AK030254.1 | 15 | 1983 | ||
| AY618264.1 | 14 | 1795 | AAT68665.1 | |
| BC066782.1 | 15 | 1847 | AAH66782.1 | |
| CK789086.1 | 4 | 359 |
More Information
Set Membership: |
|

Genomic Map