Assay Details
Target Gene Details
Entrez Gene ID: | 16963 |
Gene Name: | chemokine (C motif) ligand 1 |
Gene Aliases: |
AI661682, ATAC, LTN, Lptn, SCM-1, SCM-1a, Scyc1 |
Location: |
Chr.1:164931648-164935510 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xcl1 | NM_008510.1 | NP_032536.1 | ||
| AK079820.1 | BAC37756.1 | |||
| BC062249.1 | AAH62249.1 | |||
| D43769.1 | BAA07826.1 | |||
| U15607.1 | AAA56752.1 |
More Information
Set Membership: |
|

Genomic Map