Assay Details
Target Gene Details
Entrez Gene ID: | 77264 |
Gene Name: | zinc finger protein 142 |
Gene Aliases: |
9330177B18Rik, BB154236, mKIAA0236 |
Location: |
Chr.1:74565128-74588211 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp142 | NM_029888.2 | 6 | 2152 | NP_084164.1 |
| AB093218.1 | 6 | 2439 | BAC41402.1 | |
| AF332091.1 | 5 | 1440 | AAK56117.1 | |
| AF332092.1 | 5 | 1440 | AAK56118.1 | |
| AK132550.1 | 6 | 2153 | BAE21230.1 | |
| BC060118.1 | 6 | 2088 | AAH60118.1 |
More Information
Set Membership: |
|

Genomic Map