Assay Details
Target Gene Details
Entrez Gene ID: | 226442 |
Gene Name: | zinc finger protein 281 |
Gene Aliases: |
Znf281 |
Location: |
Chr.1:136624901-136630391 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp281 | NM_001160251.1 | 1 | 66 | NP_001153723.1 |
| NM_177643.4 | 1 | 66 | NP_808311.1 | |
| AK142218.1 | BAE24981.1 | |||
| AK157189.1 | BAE33992.1 | |||
| AV581959.1 | 1 | 99 | ||
| BC003243.1 | AAH03243.1 | |||
| BC062153.1 | AAH62153.1 |
More Information
Set Membership: |
|

Genomic Map