Assay Details
Target Gene Details
Entrez Gene ID: | 69368 |
Gene Name: | WD repeat and FYVE domain containing 1 |
Gene Aliases: |
1700013B03Rik, 1700120F24Rik, FENS-1, Jr1, WDF1, ZFYVE17, mKIAA1435 |
Location: |
Chr.1:79702262-79776047 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdfy1 | NM_001111279.1 | NP_001104749.1 | ||
| AK007215.1 | ||||
| AK157204.1 | ||||
| AK159710.1 | ||||
| AK165554.1 | ||||
| AK193130.1 | ||||
| AK219012.1 | ||||
| AK220391.1 | BAD90445.1 | |||
| BC025226.1 | AAH25226.1 |

Genomic Map