Assay Details
Target Gene Details
Entrez Gene ID: | 226757 |
Gene Name: | WD repeat domain 26 |
Gene Aliases: |
1600024A01Rik, AA693241, AI447817, AU044014, C77982, Gid7 |
Location: |
Chr.1:181173226-181211978 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdr26 | NM_145514.5 | NP_663489.4 | ||
| AK075743.1 | BAC35923.1 | |||
| AK082326.1 | ||||
| AK184747.1 | ||||
| AK187222.1 | ||||
| AK204474.1 | ||||
| AK205717.1 | ||||
| AK214736.1 | ||||
| AK219748.1 | ||||
| BC058601.1 | AAH58601.1 | |||
| BC139336.1 | ||||
| BC139337.1 |

Genomic Map