Assay Details
Target Gene Details
Entrez Gene ID: | 227326 |
Gene Name: | G protein-coupled receptor 55 |
Gene Aliases: |
Gm218, Lpir1 |
Location: |
Chr.1:85933206-85991567 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpr55 | NM_001033290.2 | 1 | 113 | NP_001028462.2 |
| AK146484.1 | 1 | 66 | BAE27205.1 | |
| BY089906.1 | 1 | 113 |
More Information
Set Membership: |
|

Genomic Map