Assay Details
Target Gene Details
Entrez Gene ID: | 52846 |
Gene Name: | CCR4-NOT transcription complex, subunit 11 |
Gene Aliases: |
2410015L18Rik, C40, D1Bwg0212e |
Location: |
Chr.1:39535802-39546881 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnot11 | NM_028043.2 | 5 | 1154 | NP_082319.1 |
| AK010503.1 | 5 | 1166 | BAB26988.1 | |
| AK041342.1 | 3 | 479 | BAC30912.1 | |
| AK144692.1 | 5 | 1169 | BAE26018.1 | |
| AK167440.1 | 5 | 1145 | BAE39526.1 | |
| BC049091.1 | 5 | 1154 | AAH49091.1 | |
| BC050812.1 | 5 | 1134 | AAH50812.1 |

Genomic Map