Assay Details
Target Gene Details
Entrez Gene ID: | 16153 |
Gene Name: | interleukin 10 |
Gene Aliases: |
CSIF, Il-10 |
Location: |
Chr.1:131019845-131024970 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il10 | NM_010548.2 | 1 | 74 | NP_034678.1 |
| AK152344.1 | 1 | 75 | BAE31139.1 | |
| BC120612.1 | ||||
| BC137844.1 | ||||
| M37897.1 | 1 | 82 | AAA39274.1 |

Genomic Map