Assay Details
Target Gene Details
Entrez Gene ID: | 12227 |
Gene Name: | B cell translocation gene 2, anti-proliferative |
Gene Aliases: |
AA959598, APRO1, Pc3, TIS21 |
Location: |
Chr.1:134074865-134079155 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Btg2 | NM_007570.2 | 1 | 120 | NP_031596.1 |
| AK088976.1 | 1 | 83 | BAC40682.1 | |
| AK133796.1 | 1 | 85 | BAE21847.1 | |
| AK169269.1 | 1 | 85 | BAE41030.1 | |
| AK169436.1 | 1 | 85 | BAE41177.1 | |
| BC132259.1 | 1 | 70 | ||
| BC138639.1 | 1 | 70 | ||
| BY766758.1 | 1 | 125 |

Genomic Map