Assay Details
Target Gene Details
Entrez Gene ID: | 12628 |
Gene Name: | complement component factor h |
Gene Aliases: |
Mud-1, NOM, Sas-1, Sas1 |
Location: |
Chr.1:140085855-140183411 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfh | NM_009888.3 | 1 | 166 | NP_034018.2 |
| AK052688.1 | 1 | 166 | ||
| AK087707.1 | 1 | 75 | ||
| BB661130.1 | 1 | 166 | ||
| BC066092.1 | 1 | 35 | AAH66092.1 | |
| J02891.1 | 1 | 519 | AAA37795.1 | |
| M12660.1 | 1 | 77 | AAA37759.1 |

Genomic Map