Assay Details
Target Gene Details
Entrez Gene ID: | 75619 |
Gene Name: | FAST kinase domains 2 |
Gene Aliases: |
2810421I24Rik |
Location: |
Chr.1:63730599-63753385 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fastkd2 | NM_172422.3 | NP_766010.1 | ||
| AK035322.1 | ||||
| AK084971.1 | BAC39325.1 | |||
| AK129251.1 | BAC98061.1 | |||
| AK153916.1 | BAE32254.1 | |||
| AK166378.1 | BAE38740.1 | |||
| AK167714.1 | ||||
| BC008271.1 | AAH08271.2 | |||
| BC057208.1 | AAH57208.1 | |||
| BC080813.1 | AAH80813.1 |

Genomic Map