Assay Details
Target Gene Details
Entrez Gene ID: | 21367 |
Gene Name: | contactin 2 |
Gene Aliases: |
D130012K04Rik, TAG-1, Tax |
Location: |
Chr.1:132509425-132543251 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cntn2 | NM_177129.5 | 18 | 2551 | NP_796103.2 |
| AK147573.1 | ||||
| AK147602.1 | 18 | 2552 | BAE28018.1 | |
| AK147658.1 | ||||
| AK147673.1 | 17 | 2704 | BAE28065.1 | |
| BC053033.1 | 7 | 759 | AAH53033.1 | |
| BC066106.1 | 18 | 2471 | AAH66106.1 | |
| X81365.1 | 3 | 259 | CAA57130.1 |
More Information
Set Membership: |
|

Genomic Map