Assay Details
Target Gene Details
Entrez Gene ID: | 21367 |
Gene Name: | contactin 2 |
Gene Aliases: |
D130012K04Rik, TAG-1, Tax |
Location: |
Chr.1:132509425-132543251 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cntn2 | NM_177129.5 | 6 | 900 | NP_796103.2 |
| AK147573.1 | 5 | 1028 | ||
| AK147602.1 | 6 | 901 | BAE28018.1 | |
| AK147658.1 | 5 | 1028 | ||
| AK147673.1 | 5 | 1053 | BAE28065.1 | |
| BC066106.1 | 6 | 820 | AAH66106.1 |
More Information
Set Membership: |
|

Genomic Map