Assay Details
Target Gene Details
Entrez Gene ID: | 241062 |
Gene Name: | post-GPI attachment to proteins 1 |
Gene Aliases: |
5033403E17Rik, 9030223K07Rik, A530084K22, D230012E17Rik, oto |
Location: |
Chr.1:54473000-54557684 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pgap1 | NM_001163314.2 | 1 | 129 | NP_001156786.1 |
| AK041133.1 | 1 | 130 | ||
| AK051360.1 | 1 | 73 | BAC34614.1 | |
| AK051866.1 | 1 | 140 | BAC34791.1 | |
| AK138171.1 | 1 | 69 | BAE23568.1 |
More Information
Set Membership: |
|

Genomic Map