Assay Details
Target Gene Details
Entrez Gene ID: | 19243 |
Gene Name: | protein tyrosine phosphatase 4a1 |
Gene Aliases: |
AA415290, AU019864, C130021B01, Prl-1 |
Location: |
Chr.1:30940303-30949765 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptp4a1 | NM_011200.2 | NP_035330.1 | ||
| AK148288.1 | BAE28460.1 | |||
| BC055039.1 | AAH55039.1 | |||
| CA315780.1 | ||||
| U84411.1 | AAB58913.1 |
More Information
Set Membership: |
|

Genomic Map