Assay Details
Target Gene Details
Entrez Gene ID: | 19264 |
Gene Name: | protein tyrosine phosphatase, receptor type, C |
Gene Aliases: |
B220, CD45R, Cd45, L-CA, Ly-5, Lyt-4, T200, loc |
Location: |
Chr.1:138062859-138175755 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprc | NM_001111316.2 | 8 | 713 | NP_001104786.2 |
| NM_001268286.1 | 5 | 322 | NP_001255215.1 | |
| NM_011210.4 | 6 | 394 | NP_035340.3 | |
| AK054056.1 | 6 | 395 | BAC35638.2 | |
| AK088875.1 | 1 | 1759 | ||
| AK154893.1 | 5 | 317 | BAE32907.1 | |
| BC028512.1 | 6 | 334 | ||
| M11934.1 | 5 | 224 | AAA39461.1 | |
| M14342.1 | 6 | 356 | AAA39458.1 | |
| M14343.1 | 2 | 887 | ||
| M17320.1 | AAA60449.1 | |||
| M92933.1 | 8 | 713 | AAA39459.1 |
More Information
Set Membership: |
|

Genomic Map