Assay Details
Target Gene Details
Entrez Gene ID: | 171531 |
Gene Name: | melanophilin |
Gene Aliases: |
2210418F23Rik, 5031433I09Rik, AW228792, D1Wsu84e, Slac-2a, l(1)-3Rk, l1Rk3, ln |
Location: |
Chr.1:90915022-90951142 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mlph | NM_053015.3 | 7 | 954 | NP_443748.2 |
| AB057759.1 | 6 | 776 | BAB41087.1 | |
| AF384098.1 | 7 | 951 | AAK97435.1 | |
| AK019883.1 | 7 | 925 | ||
| AK148274.1 | 7 | 926 | BAE28452.1 | |
| AK187519.1 | 4 | 347 | ||
| BC150804.1 | 7 | 848 |
More Information
Set Membership: |
|

Genomic Map