Assay Details
Target Gene Details
Entrez Gene ID: | 66404 |
Gene Name: | replication termination factor 2 domain containing 1 |
Gene Aliases: |
1700067C04Rik, 2410001C21Rik, 5730427M17Rik, AA589417 |
Location: |
Chr.2:172440578-172469899 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtfdc1 | NM_025542.2 | NP_079818.1 | ||
| AK009901.1 | BAB26573.1 | |||
| AK010321.1 | ||||
| AK028792.1 | BAC26123.1 | |||
| AK053258.1 | BAC35324.1 | |||
| AK077823.1 | BAC37024.1 | |||
| AK153121.1 | BAE31736.1 | |||
| AK167287.1 | BAE39393.1 | |||
| AK169588.1 | BAE41244.1 | |||
| AK170141.1 | BAE41592.1 | |||
| BC004803.1 | AAH04803.1 | |||
| BC049072.1 | AAH49072.2 | |||
| BC096692.1 | AAH96692.1 | |||
| BC116442.1 | ||||
| BC116443.1 | ||||
| BC127166.1 |

Genomic Map