Assay Details
Target Gene Details
Entrez Gene ID: | 66748 |
Gene Name: | glutamate rich 2 |
Gene Aliases: |
4933404M02Rik |
Location: |
Chr.2:70508630-70540885 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erich2 | NM_025744.2 | NP_080020.2 | ||
| AK006384.1 | ||||
| AK016654.1 | BAB30362.1 | |||
| AK017113.1 |
Target Gene Details
Entrez Gene ID: | 68986 |
Gene Name: | glutamate decarboxylase 1, opposite strand |
Gene Aliases: |
1500002O10Rik |
Location: |
Chr.2:70489940-70563357 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gad1os | NR_040496.1 | |||
| AK005124.1 |
More Information
Set Membership: |
|

Genomic Map