Assay Details
Target Gene Details
Entrez Gene ID: | 76161 |
Gene Name: | lysosomal-associated membrane protein family, member 5 |
Gene Aliases: |
3110035N03Rik, 6330527O06Rik, BAD-LAMP, LAMP-5 |
Location: |
Chr.2:136051959-136069917 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamp5 | NM_029530.2 | 2 | 495 | NP_083806.2 |
| AK018222.1 | 2 | 194 | ||
| AK045946.1 | 2 | 170 | ||
| CX240608.1 | 2 | 495 |
More Information
Set Membership: |
|

Genomic Map