Assay Details
Target Gene Details
Entrez Gene ID: | 60525 |
Gene Name: | acyl-CoA synthetase short-chain family member 2 |
Gene Aliases: |
1110017C11Rik, ACAS, Acas1, Acas2, AceCS1, Acs1 |
Location: |
Chr.2:155517953-155562746 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acss2 | NM_019811.3 | NP_062785.2 | ||
| AF216873.1 | AAF24510.1 | |||
| AK028586.1 | BAC26019.1 | |||
| AK029014.1 | BAC26243.1 | |||
| AK053877.1 | BAC35571.1 | |||
| AK086921.1 | 3 | 1439 | ||
| AK093831.1 | BAC04235.1 | |||
| BC051432.1 | AAH51432.1 |
Target Gene Details
Entrez Gene ID: | 74945 |
Gene Name: | acyl-CoA synthetase short-chain family member 2, opposite strand |
Gene Aliases: |
4930471I20Rik |
Location: |
Chr.2:155547040-155556930 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acss2os | NR_040613.1 | |||
| AK015552.1 | ||||
| BY715100.1 |
More Information
Set Membership: |
|

Genomic Map