Assay Details
Target Gene Details
Entrez Gene ID: | 109275 |
Gene Name: | ARP5 actin-related protein 5 |
Gene Aliases: |
AA545173, B430109J19Rik |
Location: |
Chr.2:158624638-158639211 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actr5 | NM_175419.4 | NP_780628.3 | ||
| AK046312.1 | ||||
| AK142980.1 |
More Information
Set Membership: |
|

Genomic Map